missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Microtube Cutter
Safely, accurately and easily sections microcentrifuge tubes and eliminates the hazards of using razor blades or knives
Brand: Bel-Art™ F21018-0000
Code : 50
Additional Details : Weight : 0.07200kg
Description
- Makes it easy to retrieve radioactive pellets from the tips of tubes without resuspension or the risk of splashed radioactive substances
- Works well with polyethylene c or polypropylene microcentrifuge tubes up to 1.5mL
- Chrome-plated body with replaceable steel blade
- Maximum cut diameter is 9.4mm (0.370 in.)
Not recommended for polystyrene tubes.
Specifications
Microtube Cutter | |
Chrome-plated Steel | |
Microtube cutter |