missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Microcentrifuge Tube Sample Pestles

Designed to homogenize samples in a microcentrifuge tube

Brand:  Bel-Art™ F19922-0001

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11782324

  • £71.40 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Suitable for resuspending lyophilized products or precipitates
  • fluoropolymer pestle is mounted on a stainless steel shaft, 3.2mm (0.125 in.) in diameter
  • May be hand-held or mounted in a motorized chuck
  • Overall length 14.3cm (5.625 in.)
  • Steam autoclavable at 121°C (250°F)
  • Choose rounded or flat tip shape to match centrifuge type

Specifications

Specifications

1.5mL Microcentrifuge tube
143mm
Designed to homogenize samples in a microcentrifuge tube
Teflon™ PTFE pestle, Stainless Steel shaft
Yes
Videos