missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Micro Funnels

Designed for micro work, filling burets, manometers and small bottles

Brand:  Bel-Art™ F14685-0028

Code : 02

Additional Details : Weight : 0.02068kg

 View more versions of this product

Product Code. 12314016

  • £27.30 / Pack of 12


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Polypropylene funnels are steam autoclavable at 121°C (250°F)

Specifications

Specifications

Yes
For micro work, filling burettes, manometers and small bottles
PP (Polypropylene)
0.6cm
5.2mL
Videos