Learn More
Bel Art Products Colorimeter, Industrial; Bel-Art Scienceware; KLETT; Model: 900-3; For colorimetric/turbidimetric analyses; Accessory filters; Photovoltaic cells; Suspension galvanometer; w/Blue (400-450nm)/green (520-580nm) glass filters; 220VAC
Brand:  Bel Art Products T37013-0001
Colorimeter, Industrial; Bel-Art Scienceware; KLETT; Model: 900-3; For colorimetric/turbidimetric analyses; Accessory filters; Photovoltaic cells; Suspension galvanometer; w/Blue (400-450nm)/green (520-580nm) glass filters; 220VAC
Code : 03
Additional Details : Weight : 5.70000kg
Product Code. 15346513