missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Earth Friendly 1001 Sticks

Lessen environmental impact with polylactic acid (PLA) resin sampling sticks

Brand:  Bel-Art™ F36931-0000

Code : 04

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 13402489

  • £39.29 / Pack of 100


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

The production of 100% plant-based PLA contributes fewer greenhouse gases and uses less nonrenewable energy from source to resin than its petroleum-based counterparts, and the plants used are an annually renewable resource.

  • Suitable for mixing samples, handling capsules, scraping containers or positioning solid samples for examination
  • Green alternative to petroleum-based sampling tools helps meet sustainability goals
  • Comparable in strength to polystyrene, polypropylene and other petroleum-based plastic resin sampling tools
  • Disposal options include industrial composting for biodegradability, incineration, recycling and landfill
  • Nonsterile FDA-grade: safe for food, drug and cosmetic contact
  • Use a separate stick for each material sampled to avoid cross-contamination

Specifications

Specifications

100/Pk.
Earth-Friendly™ Sampling Stick
No
5.75 x 0.75 in. (14.6 x 1.9cm)
14.6cm
FDA
Suitable for mixing samples, handling capsules, scraping containers or positioning solid samples for examination
1.9cm
PLA Resin
Videos