missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Autoradiography Pens

Easy marking of film with nontoxic and nonradioactive phosphorescent material

Brand:  Bel-Art™ F13350-0000

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12320469

  • £88.98 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Includes: Pocket clip and screw-knob advance to adjust writing tip.

Description

Description

  • Solid phosphorescent material will not leak or dry out, (no bladder, vial or liquids to spill) Nonradioactive energy may be adjusted by exposure to white incandescent or fluorescent light
  • 3 to 4 minute energy emission leaves distinct impression without overexposing film
  • Bright green markings do not glow in the dark but are visible under normal conditions and on X-ray film
  • No Beta or other isotope emissions; Spectral peak, 5600Å
  • Screw knob advances writing tip; Pens are 4 1/2 in. long (11.4cm)
  • Not appropriate for phosphor screens

High Energy Level Markers
Greater energy emission strength and slightly longer energy emission duration
Suitable for marking X-Ray films similar to Kodak™ XLS
No Beta or other isotope emissions
Marking is unaffected by chilled films and an intensifier screen will not affect intensity
Not appropriate for phosphor screens

Specifications

Specifications

Ideal for marking various films that are to be X-rayed with serial numbers, dates and exposure periods, film position markings
114.3mm
Each
Pocket clip and screw-knob advance to adjust writing tip
Non Radioactive Marker
Bright Green
Videos