Bel-Art™ SP Scienceware™ Antibody Saver Trays

Minimize the use of expensive antibodies during incubations by using an optimally sized tray

Brand: Bel-Art™

Manufacturer Part Number: F45100-0001

X5 Antibody Saver Trays, 9 x 9 cm, polypropylene,autoclavable

UNSPSC: 41100000

Code: Z4

Additional Details:
Additional Details: Weight: 11.34000kg

Product Code. 15289722

Quantity Price
1 £110.14 / Pack of 5
Call for latest availability
+44 (0)1509 555 500
Add to basket

Description and Specification


Length (Metric) 9cm
Width (Metric) 9cm
Autoclavable Yes
Type Antibody Saver Tray
Material Polypropylene
Includes Attached lid to protect against spillage
Dimensions (L x W) 3.5 x 3.5 in. (9 x 9cm)

  • For popular midi and mini gels
  • Stackable, reclosable and reusable
  • All trays include attached lid to protect against spillage
  • Single lane trays are supplied as a group of five joined lanes; can be easily separated to single lane prior to use
  • Polypropylene; autoclavable