missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Antibody Saver Trays
Minimize the use of expensive antibodies during incubations by using an optimally sized tray
£41.60
Specifications
Material | Polypropylene |
---|---|
Type | Antibody Saver Tray |
Includes | Attached lid to protect against spillage |
Autoclavable | Autoclavable |
Product Code | Brand | Autoclavable | Length (English) | Width (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Autoclavable | Length (English) | Width (English) | Price | Quantity & Availability | |||
12860713
|
Bel-Art™
F45100-0000 |
Autoclavable | 0.625 in. | 4.5 in. |
£41.60
Pack of 5 |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
Description
- For popular midi and mini gels
- Stackable, reclosable and reusable
- All trays include attached lid to protect against spillage
- Single lane trays are supplied as a group of five joined lanes; can be easily separated to single lane prior to use
- Polypropylene; autoclavable
Specifications
Polypropylene | |
Antibody Saver Tray | |
Attached lid to protect against spillage | |
Autoclavable |