Learn More
Bel-Art™ SP Scienceware™ Vortexing Attachment for PrepSafe™ Microcentrifuge Tube Mini Floating Racks
For samples to be vortexed simultaneously
Brand: Bel-Art™ F18742-0140
Code : NEW
Additional Details : Weight : 0.08570kg
Specifications
Vortexing Attachment | |
Natural | |
Polypropylene | |
For samples to be vortexed simultaneously | |
1 Each |
Autoclavable | |
Tubes (1.5 to 2 mL) | |
1 | |
Microcentrifuge tube mini floating racks |