missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beclomethasone dipropionate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 208.0°C |
Loss on Drying | 1% max. (105°C, 3 h) |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (HPLC) |
Quantity | 250 mg |
Formula Weight | 521.04 |
Percent Purity | 97% |
Chemical Name or Material | Beclomethasone dipropionate |
Safety and Handling
Product Identifier
- Beclomethasone dipropionate
- Danger
- Reproductive toxicity Category 1B
- H360-May damage fertility or the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'