missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BDCS, silylation reagent, AcroSeal™, Thermo Scientific™
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Color | Yellow |
Density | 0.9590g/mL |
Flash Point | 58°C |
Infrared Spectrum | Authentic |
Packaging | AcroSeal™ Glass bottle |
Refractive Index | 1.4400 to 1.4420 |
Quantity | 100 mL |
Fieser | 07,184; 01,489; 03,157; 08,255 |
Specific Gravity | 0.959 |
Formula Weight | 218.8 |
Show More |
Safety and Handling
Product Identifier
- BDCS
- Danger
- Acute toxicity Category 4
- Flammable liquid Category 3
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H361d-Suspected of damaging the unborn child.
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
- EUH029-Contact with water liberates toxic gas.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'