Learn More
BD Stemflow™ Human Pluripotent Stem Cell Sorting and Analysis Kit
Analyzes populations of live and fixed cells for expression of pluripotency markers and differentiation markers and to sort live populations of pluripotent stem cells and their derivatives
Brand: BD 560461
Additional Details : Weight : 0.15000kg
Alert:
Reagents in this kit contain sodium azide. Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.
Includes: FITC SSEA-1, PE SSEA-3, Alexa Fluor™ 647 TRA-1-81, FITC isotype control, PE isotype control, Alexa Fluor 647 isotype control, Anti-mouse beads, Anti-rat beads, Negative beads
Description
This kit provides reagents necessary to perform multicolor flow cytometry on human pluripotent stem cells such as human embryonic stem cells (hESCs) and induced pluripotent stem (iPS) cells.
- Reactivity: human
- Size: 50 tests
Specifications
FITC SSEA-1, PE SSEA-3, Alexa Fluor™ 647 TRA-1-81, FITC isotype control, PE isotype control, Alexa Fluor™ 647 isotype control, Anti-mouse beads, Anti-Rat beads, negative beads | |
50 Tests |