Learn More
BD H-2Ld:Ig Recombinant Soluble Dimeric Fusion Protein, Mouse
Consists of three extracellular major histocompatibility complex (MHC) class I H-2Ld domains that are fused to the VH regions of mouse IgG1, supplemented with recombinant β2M
Brand: BD 550751
Additional Details : Weight : 0.01000kg
Description
- Isotype: Mouse IgG1, λ
- Storage buffer: Aqueous buffered solution containing ≤0.09% sodium azide
Peptide Loading, Immunofluorescent staining, flow cytometric analysis
Specifications
0.5mg/mL | |
Aqueous Buffered solution containing 0.09% Sodium Azide | |
7.2 | |
0.25mg | |
Recombinant |
Flow Cytometry, Immunofluorescent Staining, Peptide Loading | |
Mouse H-2Ld:Ig Recombinant Fusion Protein | |
Affinity Chromatography, SDS-PAGE | |
4°C | |
Solution |