missing translation for 'onlineSavingsMsg'
Learn More

BD™ GasPak™ Supplies

Used with the GasPak™ EZ Gas Generating Systems. BD™ GasPak™ Supplies are used with these complete, integrated systems for rapid, waterless and catalyst-free gas generation. Can be used to support an anaerobic, microaerophilic or CO2 enriched environment.

Brand:  BD™ ANF-404-050B

Code : 03

Additional Details : Weight : 0.04700kg

 View more versions of this product

Product Code. 12370439

  • £55.79 / Pack of 10


Estimated Shipment
06-06-2018
Add to basket

Description

Description

GasPak™ EZ Gas Generating Systems are complete, integrated systems that rapidly produce atmospheres suitable to support the primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets inside multi-use incubation containers.

Accessories include:

  • CO2 Indicator Strips
  • Dry Anaerobic Indicator Strips
  • Tube Holder
  • Catalyst Replacement Charges
  • Catalyst Reaction Chamber
Specifications

Specifications

Catalyst Charges
Pack of 10
Produce atmospheres suitable to support the primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria
Catalyst Replacement Charges
Replacement charges
15° to 30°C
BD GasPak™
Videos
Documentation

Technical Documents