missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Antibiotic Medium 1
For determining antibiotic potency by the microbiological assay technique. BD Difco™ Dehydrated Culture Media: Antibiotic Medium 1 is part of a large selection of dehydrated media.
Brand: BD 226340
Additional Details : Weight : 0.52200kg
Description
- Penassay Seed Agar Microbiological assay of antibiotics
Specifications
AOAC, EP, USP | |
Dehydrated | |
Antibiotic Medium | |
Penassay Seed Agar Microbiological assay of Antibiotics | |
500 g |
2°C to 8°C | |
Antibiotic Medium 1 | |
Beef extract 1.5g/L, Yeast extract 3g/L, Pancreatic digest of Casein 4g/L, Peptone 6g/L, Dextrose 1g/L, Agar 15g/L | |
Bottle |