Learn More
BD Cytometric Bead Array (CBA) Human Enhanced Sensitivity Master Buffer Kit
Employs particles with discrete fluorescence intensities to detect soluble analytes at very low concentrations
£515.00 - £1565.00
Specifications
Cross Reactivity | Human |
---|---|
Includes | Assay diluent, capture bead Diluent, detection reagent diluent, Wash Buffer, enhanced sensitivity detection reagent (Part B), instrument setup beads (A1, A9, F1, F9), PE instrument setup bead F1, PE positive control detector |
Content And Storage | 2°C to 8°C |
For Use With (Application) | Flow Cytometry |
Product Type | Human enhanced sensitivity master Buffer kit |
Includes: Assay Diluent, Capture Bead Diluent, Detection Reagent Diluent, Wash Buffer, Enhanced Sensitivity Detection Reagent (Part B), Instrument Setup Beads (A1, A9, F1, F9), PE Instrument Setup Bead F1, PE Positive Control Detector
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15851099
|
BD
561521 |
100 Tests |
£515.00
Each |
Estimated Shipment: 26-04-2024
Log in to see stock. |
|||||
15871099
|
BD
561523 |
500 Tests |
£1565.00
Each |
Estimated Shipment: 26-04-2024
Log in to see stock. |
|||||
Description
Detection reagents used in BD™ CBA Human Enhanced Sensitivity Flex Set System consist of two parts, Analyte Specific Detection Reagent (Part A), provided with the Flex Set and Enhanced Sensitivity Detection Reagent (Part B), provided in Master Buffer Kit with all other supporting reagents. Buffers, detection reagent and instrument setup reagents provided in this kit have been optimized for analysis of analytes in tissue culture supernatants, plasma, and serum samples.
- Analyte working assay range: 274 to 200000fg/mL
- Should not be used with Flex Sets from any other BD CBA System such as soluble protein, cell signalling etc
- Available in pack sizes of 100 and 500 tests
Flow Cytometry
Specifications
Human | |
2°C to 8°C | |
Human enhanced sensitivity master Buffer kit |
Assay diluent, capture bead Diluent, detection reagent diluent, Wash Buffer, enhanced sensitivity detection reagent (Part B), instrument setup beads (A1, A9, F1, F9), PE instrument setup bead F1, PE positive control detector | |
Flow Cytometry |