Learn More
BD Cytometric Bead Array (CBA) Human CD62E (E-Selectin); Flex Set, Bead D9
Bead-based immunoassay capable of measuring human soluble CD62E (E-Selectin) in serum, plasma and cell culture supernatant samples
Brand: BD 560419
Additional Details : Weight : 0.04430kg
Includes:
- Human soluble CD62E (E-Selectin), Standard
- Human soluble CD62E (E-Selectin), PE Detection Reagent
- Human soluble CD62E (E-Selectin), Capture Bead D9
Description
Contains one vial each of Capture Bead and PE Detection Reagent and two vials of Standard. The Capture Bead and PE Detection Reagent components of this flex set have been formulated to a 50x concentration to ensure product performance when multiplexed. The Standard component is lyophilized and should be transferred to a 15mL polypropylene tube for reconstitution. When reconstituted in 4.0mL Assay Diluent, the standard has a protein concentration of 10,000 pg/mL.
- Human reactivity was determined by testing samples with CBA Human Soluble CD62E (E-Selectin) Flex Set
- Must be used in conjunction with CBA Human Soluble Protein Master Buffer Kit, flow cytometer, and FCAP Array Software
Specifications
CD62E (E-Selectin) | |
Human soluble CD62E (E-Selectin), standard, Human soluble CD62E (E-Selectin), PE detection reagent, Human soluble CD62E (E-Selectin), capture bead D9 | |
4°C | |
Human CD62E (E-Selectin) flex set (bead D9) |
Human | |
100 Tests | |
Flow Cytometry |