missing translation for 'onlineSavingsMsg'
Learn More

BD Cytofix/Cytoperm™ Fixation/Permeabilization Solution Kit

Enables the fixation and permeabilization of cells which is necessary for staining intracellular cytokines with fluorochrome-conjugated anti-cytokine antibodies

Brand:  BD 554714

Code : NEW

Additional Details : Weight : 0.00100kg


Product Code. 15747847

  • £446.00 / 250 tests

In Stock 1

Please to purchase this item. Need a web account? Register with us today!


You may only order up to the available stock. For more details call customer services on 01509 555 500

Includes:
  • Fixation/Permeabilization solution (125mL)
  • Perm/Wash Buffer, 10x concentrate containing Fetal Bovine Serum (FBS) and saponin (100mL)

Description

Description

  • Provides two reagents, fixation/permeabilization solution and Perm/Wash Buffer
  • After cell fixation and permeabilization, the Perm/Wash Buffer is used to wash the cells and to dilute the anti-cytokine antibodies for staining

Specifications

Specifications

Fixation/Permeabilization Solution Kit
For staining intracellular cytokines with fluorochrome-conjugated Anti-cytokine antibodies
125mL fixation/permeabilization solution, perm/Wash™ buffer, fetal bovine serum (FBS) 10X and 100mL saponin
4°C
RUO
Flow cytometer
250.0
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BD Cytofix/Cytoperm Fixation/Permeabilization Solution Kit

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B

Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Formaldehyde

Documents