Learn More
BD Pharmingen™ Calcein Blue AM Fluorescent Dye
Product Code. 15803329
Click to view available options
Quantity:
1 mg
Unit Size:
Each
Product Code. 15803329
Brand: BD 564060
Calcein Blue AM is used for labeling live cells that can be detected and further analyzed by flow cytometry.
  • Fluoresent dye for labeling live cells.
TRUSTED_SUSTAINABILITY
Specifications
Description Calcein AM fluorescent dye
For Use With (Application) Flow Cytometry
Quantity 1 mg