missing translation for 'onlineSavingsMsg'
Learn More

BD 1063 dihydrochloride, Tocris Bioscience™

Selective σ1 ligand, putative antagonist

Brand:  Tocris (Bio-Techne) 088310

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13488103

  • £127.00 / 10mg


Estimated Shipment
07-03-2018
Add to basket

Description

Description

σ1 receptor antagonist, approximately 50-fold selective over σ2 sites and ≥100-fold selective over opioid, PCP, muscarinic, dopamine, α1, α2, β adrenoceptor, 5-HT1 and 5-HT2 receptors. Prevents hyperlocomotion following cocaine administration in mice.
Specifications

Chemical Identifiers

BD 1063 dihydrochloride
C13H18Cl2N2·2HCl
346.13
1-[2-(3, 4-Dichlorophenyl)ethyl]-4-methylpiperazine dihydrochloride

Specifications

BD 1063 dihydrochloride
206996-13-6
C13H18Cl2N2·2HCl
1-[2-(3, 4-Dichlorophenyl)ethyl]-4-methylpiperazine dihydrochloride
10mg
>99%
346.13
Videos