Learn More
BD 1047 dihydrobromide, Tocris Bioscience™
σ1 selective antagonist
Brand:  Tocris (Bio-Techne) 0956/50
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13438203
  • £521.85 / 50mg
Estimated Shipment date
from Supplier 25-10-2019
Description
Description
σ1 receptor antagonist, exhibiting a similar binding profile to that of BD 1063 (Cat. No. 0883) but with higher affinity (approximately 10-fold) at both σ1 and σ2 sites.
Specifications
Chemical Identifiers
138356-20-4
275.217
unii-1s3x75qgdo, 1s3x75qgdo, n-2-3,4-dichlorophenyl ethyl-n-methyl-2-dimethylamino ethylamine, n-2-3,4-dichlorophenyl ethyl-n,n',n'-trimethylethane-1,2-diamine, 2-3,4-dichlorophenyl ethyl 2-dimethylamino ethyl methylamine, tocris-0956, d09cgh, n-2-3,4-dichlorphenyl ethyl-n,n',n'-trimethyl-1,2-ethandiamin, n1-3,4-dichlorophenethyl-n1,n2,n2-trimethylethane-1,2-diamine, 1,2-ethanediamine, n-2-3,4-dichlorophenyl ethyl-n,n',n'-trimethyl
N'-[2-(3,4-dichlorophenyl)ethyl]-N,N,N'-trimethylethane-1,2-diamine
C13H20Cl2N2
MGVRNMUKTZOQOW-UHFFFAOYSA-N
188914
CN(C)CCN(C)CCC1=CC(=C(C=C1)Cl)Cl
Specifications
50mg
138356-20-4
unii-1s3x75qgdo, 1s3x75qgdo, n-2-3,4-dichlorophenyl ethyl-n-methyl-2-dimethylamino ethylamine, n-2-3,4-dichlorophenyl ethyl-n,n',n'-trimethylethane-1,2-diamine, 2-3,4-dichlorophenyl ethyl 2-dimethylamino ethyl methylamine, tocris-0956, d09cgh, n-2-3,4-dichlorphenyl ethyl-n,n',n'-trimethyl-1,2-ethandiamin, n1-3,4-dichlorophenethyl-n1,n2,n2-trimethylethane-1,2-diamine, 1,2-ethanediamine, n-2-3,4-dichlorophenyl ethyl-n,n',n'-trimethyl
CN(C)CCN(C)CCC1=CC(=C(C=C1)Cl)Cl
275.217
437.04
BD 1047 dihydrobromide
C13H20Cl2N2
MGVRNMUKTZOQOW-UHFFFAOYSA-N
N'-[2-(3,4-dichlorophenyl)ethyl]-N,N,N'-trimethylethane-1,2-diamine
188914
>99%