missing translation for 'onlineSavingsMsg'
Learn More

BD 1008 dihydrobromide, Tocris Bioscience™

Potent, selective σ ligand

Brand:  Tocris (Bio-Techne) 0511/50

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13487743

  • £506.00 / 50mg


Estimated Shipment
08-03-2018
Add to basket

Description

Description

Potent and selective σ-ligand (Ki against [3H]-(+)-3-PPP = 0.34 nM).
Specifications

Chemical Identifiers

BD 1008 dihydrobromide
C15H22Cl2N2·2HBr
463.08
N-[2-(3, 4-Dichlorophenyl)ethyl]-N-methyl-2-(1-pyrrolidinyl)ethylamine dihydrobromide

Specifications

BD 1008 dihydrobromide
138356-09-9
C15H22Cl2N2·2HBr
N-[2-(3, 4-Dichlorophenyl)ethyl]-N-methyl-2-(1-pyrrolidinyl)ethylamine dihydrobromide
50mg
>98%
463.08
Videos