missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCMA Neutralizing Mouse, Unconjugated, Monoclonal Antibody, recommended for neutralizing assay, AcroBiosystems
Mouse Monoclonal Antibody
£297.00 - £2160.00
Specifications
| Antigen | BCMA Neutralizing |
|---|---|
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Host Species | Human (HEK293) |
| Regulatory Status | RUO |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17146091
|
ACROBiosystems
BCA-M43-100ug |
100 μg |
£297.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17156091
|
ACROBiosystems
BCA-M43-1mg |
1 mg |
£2160.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BCMA Neutralizing Monoclonal antibody specifically detects BCMA Neutralizing, recommended for neutralizing assay.Specifications
| BCMA Neutralizing | |
| Unconjugated | |
| RUO | |
| Primary |
| Monoclonal | |
| Human (HEK293) | |
| IgG1 | |
| -20°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title