missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ BCL6 Recombinant Human Protein, N-Trx.His tag

Expressed from DNA sequence encoding human BCL6 N-terminal fragment (Met 1-Met 150) fused to Trx and polyhistidine tag at N-terminus

Brand:  Sino Biological™ 12083H30E50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15215745

  • £157.44 / 50µg


Estimated Shipment
11-06-2018
Add to basket

Description

Description

  • Formulation: 20mM Tris, 10% glycerol, pH 8.0
Specifications

Specifications

Used as protein standard and in cell biology research applications
604
≥90% by SDS-PAGE
Human
B-cell CLL/Lymphoma 6
Assay Standard
E.coli
Thioredoxin, His Tag
Lyophilized
34.2kd
50μg
Truncated, Recombinant
BCL6
Transcription Factor
BCL6 Recombinant Human Protein
Videos

For Research Use Only. Not for use in diagnostic procedures.