Learn More
BCL6 Monoclonal Antibody (BCL-UP), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£106.00 - £285.00
Specifications
Antigen | BCL6 |
---|---|
Clone | BCL-UP |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15568266
|
Affymetrix eBioscience
17-9880-41 |
25 Tests |
£106.00
25 tests |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15578266
|
Affymetrix eBioscience
17-9880-42 |
100 Tests |
£285.00
100 tests |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The monoclonal antibody BCL-UP recognizes Bcl-6, also known as B-cell lymphoma 6,which is a member of the POZ family of transcriptional repressors. Bcl-6 is expressed most abundantly in germinal center B cells, as well as in differentiating and activated B cells. This 79 kDa protein is essential for germinal center B cell proliferation and survival. During B cell development, Bcl-6 plays an important role in promoting pre-B cell survival to enable the generation of a diverse B cell repertoire. Moreover, Bcl-6 is expressed constitutively in diffuse large B cell lymphomas (DLBCLs), Burkitt's lymphoma, and non-Hodgkin's lymphoma. Reports also suggest expression of Bcl-6 in CD4+ T follicular helper cells and memory T cells. Bcl-6 has been reported to be antagonized by Blimp-1. Applications Reported: This BCL-UP antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This BCL-UP antibody has been pre-titrated and tested in human tonsil cells by intracellular staining with the Foxp3 Fixation/Permabilization Buffers (cat. 00-5521) and protocol. Please see Best Protocols Section (Staining intracellular Antigens for Flow Cytometry) for staining protocol (refer to Protocol B: One-step protocol for intracellular (nuclear) proteins). This can be used at 5 μL (0.125 μg) per test.
BCL6 Monoclonal antibody specifically detects BCL6 in Human samples. It is validated for Flow Cytometry.Specifications
BCL6 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
B cell CLL/lymphoma 6; B cell leukemia/lymphoma 6; B-cell CLL/lymphoma 6; B-cell CLL/lymphoma 6 (zinc finger protein 51); B-cell leukemia/lymphoma 5; B-cell leukemia/lymphoma 6; B-cell lymphoma 5 protein; B-cell lymphoma 5 protein (BCL5); B-cell lymphoma 6 protein; B-cell lymphoma 6 protein homolog; B-cell lymphoma 6 protein transcript; BCL5; BCL-5; Bcl6; Bcl-6; BCL6 transcription repressor; BCL6A; cys his2 zinc finger transcription factor; cys-his2 zinc finger transcription factor; LAZ3; LAZ-3 protein; Lymphoma Associated Zinc Finger Gene On Chromosome 3 (LAZ3); lymphoma-associated zinc finger gene on chromosome 3; protein LAZ-3; ZBTB27; zinc finger and BTB domain-containing protein 27; Zinc finger and BTB domain-containing protein 27 (ZBTB27); Zinc finger protein 51; Zinc Finger Protein 51 (ZNF51); zinc finger transcription factor BCL6S; ZNF51 | |
BCL6 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
BCL6 |
BCL-UP | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
P41182 | |
604 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.