missing translation for 'onlineSavingsMsg'
Learn More

BCL6 Mouse anti-Human, APC, Clone: BCL-UP, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-9880-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15578266

  • £228.00 / 100 tests

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. Bcl-6 proto-oncogene product (a Kruppel-type zinc-finger protein) is mainly expressed in normal germinal center B cells and related lymphomas. Bcl-6 is involved in chromosome rearrangements at 3q27 in non-Hodgkin′s lymphomas and Bcl-6 rearrangements have been detected in 33%-45% of diffuse large B cell lymphomas. Bcl-6 has been detected immunohistochemically in follicular lymphomas, diffuse large B cell lymphomas, Burkitt′s lymphomas and in nodular, lymphocyte predomit Hodgkin′s disease.
Specifications

Specifications

BCL6
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P41182
B-cell lymphoma 6 protein; BCL-6; Zinc finger protein 51; LAZ-3 protein; BCL-5; Zinc finger and BTB domain-containing protein 27; ZNF51; ZBTB27; LAZ3; BCL5; BCL6
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
BCL-UP
APC
BCL6
Liquid
BCL6
IgG1, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
604
Videos
Documents

For Research Use Only.