missing translation for 'onlineSavingsMsg'
Learn More

Bcl-2 Mouse anti-Mouse, Rat, eFluor® 660, Clone: 10C4, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-6992-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15590007

  • £65.75 / 25 tests

Estimated Shipment
10-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. BCL-2 is a key regulator of apoptosis that functions to either inhibit or promote cell death. The BCL-2 family members are also characterized by dimerizing to further modulate apoptosis. Bag1, for example, has been found to form a heterodimer with BCL-2 resulting in the enhancement of the anti-apoptotic effect of BCL-2. Bax and Bak have been shown to play a critical role in cytochrome c release from mitochondria and thus initiate apoptosis. Bax exerts a pro-apoptotic rather than an anti-apoptotic effect on cells. Constitutive expression of BCL2, such as in the case of translocation of BCL2 to Ig heavy chain locus, is thought to be the cause of follicular lymphoma. In most follicular lymphomas, neoplastic germinal centers express high levels of BCL-2 alpha protein, whereas the normal or hyperplastic germinal centers are negative. Two transcript variants of BCL-2, produced by alternate splicing, differ in their C-terminal ends. The overexpression of BCL-2 has been linked to human cancers such as B-cell lymphoma and prostate cancer.
Specifications

Specifications

Bcl-2
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P10417, P49950
AW986256, Bcl-2, C430015F12Rik, D630044D05Rik, D830018M01Rik
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse, Rat
Flow Cytometry
10C4
eFluor™ 660
Bcl2
Liquid
Bcl2
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
12043, 24224
Videos
Documents

For Research Use Only.