missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Bcl-2α Ab-3 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide
Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and flow cytometry experiments.
£105.98 - £412.48
Specifications
Antigen | Bcl-2alpha Ab-3 |
---|---|
Clone | 8C8 |
Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Host Species | Mouse |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12691707
|
Epredia™
MS-597-B0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12621717
|
Epredia™
MS-597-P0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12641717
|
Epredia™
MS-597-P1ABX |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12651717
|
Epredia™
MS-597-PABX |
Concentrated | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12601717
|
Epredia™
MS-597-B1 |
Concentrated | 500 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12631717
|
Epredia™
MS-597-P1 |
Concentrated | 500 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12611717
|
Epredia™
MS-597-P |
Concentrated | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Expression of Bcl-2α oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of Bcl-2α protein, whereas the normal or hyperplastic germinal centers are negative.Host Species: Mouse
Clone: 8C8
Isotype: IgG1
Species Reactivity: Human and Monkey. Does not react with mouse and rat. Others not tested.
Epitope: aa 41-54
Immunogen: A synthetic peptide, aa 41-54 (Cys-GAAPAPGIFSSQPG) of human Bcl-2 protein
Molecular Weight: 25-26kDa
Positive Control: Raji or HeLa Cells, Tonsil or follicular lymphomas
Cellular Localization: Cytoplasm
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
Bcl-2alpha Ab-3 | |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Mouse | |
Apoptosis | |
A Synthetic peptide, aa 41-54 (Cys-GAAPAPGIFSSQPG) of human bcl-2 protein | |
Primary |
8C8 | |
Monoclonal | |
RUO | |
Human, Non-human Primate | |
IgG1 |