Learn More
BC 11 hydrobromide, Tocris Bioscience™
Selective urokinase (uPA) inhibitor
Brand:  RD Systems (Bio-Techne) 4372/50
Code : Z1
Additional Details : Weight : 0.00050kg
Quantity 50mg
 View more versions of this product
Product Code. 13444644
  • £574.62 / 50mg
Estimated Shipment
27-06-2019
Description
Description
Selective urokinase (uPA) inhibitor (IC50 = 8.2 μM). Exhibits no activity at 8 other related enzymes. Inhibits clot lysis with no effect on clot formation. Also decreases viability of MDA-MB231 breast cancer cells in vitro.
Specifications
Chemical Identifiers
443776-49-6
290.97
bc 11 hydrobromide, carbamimidothioic acid 4-boronophenyl methyl ester hydrobromide
[4-(carbamimidoylsulfanylmethyl)phenyl]boronic acid;hydrobromide
C8H12BBrN2O2S
PAFZAMOVHIRQOD-UHFFFAOYSA-N
78243712
B(C1=CC=C(C=C1)CSC(=N)N)(O)O.Br
Specifications
BC 11 hydrobromide
443776-49-6
bc 11 hydrobromide, carbamimidothioic acid 4-boronophenyl methyl ester hydrobromide
B(C1=CC=C(C=C1)CSC(=N)N)(O)O.Br
290.97
290.97
50mg
C8H12BBrN2O2S
PAFZAMOVHIRQOD-UHFFFAOYSA-N
[4-(carbamimidoylsulfanylmethyl)phenyl]boronic acid;hydrobromide
78243712
>98%
Videos