missing translation for 'onlineSavingsMsg'
Learn More

BC 11-38, Tocris Bioscience™

Selective PDE11 inhibitor

Brand:  RD Systems (Bio-Techne) 4496/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13444844

  • £183.75 / 10mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective phosphodiesterase (PDE) 11 inhibitor (IC50 = 0.28 μM for PDE11; IC50 values are >100 μM for PDE1 - PDE10). Significantly elevates cAMP levels and cortisol production in H295R human adenocarcinoma cells.
Specifications

Chemical Identifiers

686770-80-9
304.426
d03qth, 3-phenyl-2-propylsulfanyl-6,7-dihydrothieno 3,2-d pyrimidin-4-one, 3-phenyl-2-propylsulfanyl-3h,4h,6h,7h-thieno 3,2-d pyrimidin-4-one, 3-phenyl-2-propylsulfanyl-6h,7h-thieno 3,2-d pyrimidin-4-one, 3-phenyl-2-propylthio-6,7-dihydrothieno 3,2-d pyrimidin-4 3h-one, 6,7-dihydro-3-phenyl-2-propylthio thieno 3,2-d pyrimidin-4 3h one
3-phenyl-2-propylsulfanyl-6,7-dihydrothieno[3,2-d]pyrimidin-4-one
C15H16N2OS2
YHNDCCKFNWDQGW-UHFFFAOYSA-N
2052828
CCCSC1=NC2=C(C(=O)N1C3=CC=CC=C3)SCC2

Specifications

10mg
686770-80-9
C15H16N2OS2
YHNDCCKFNWDQGW-UHFFFAOYSA-N
3-phenyl-2-propylsulfanyl-6,7-dihydrothieno[3,2-d]pyrimidin-4-one
2052828
BC 11-38
>98%
d03qth, 3-phenyl-2-propylsulfanyl-6,7-dihydrothieno 3,2-d pyrimidin-4-one, 3-phenyl-2-propylsulfanyl-3h,4h,6h,7h-thieno 3,2-d pyrimidin-4-one, 3-phenyl-2-propylsulfanyl-6h,7h-thieno 3,2-d pyrimidin-4-one, 3-phenyl-2-propylthio-6,7-dihydrothieno 3,2-d pyrimidin-4 3h-one, 6,7-dihydro-3-phenyl-2-propylthio thieno 3,2-d pyrimidin-4 3h one
CCCSC1=NC2=C(C(=O)N1C3=CC=CC=C3)SCC2
304.426
304.43
Videos
Documents