missing translation for 'onlineSavingsMsg'
Learn More

(±)-Bay K 8644, Tocris Bioscience™

CaV1.x activator

Brand:  Tocris (Bio-Techne) 154450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13459643

  • £723.24 / 50mg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

L-type Ca2+ channel activator (EC50 = 17.3 nM). Has positive inotropic, vasoconstrictive and behavioral effects in vivo. In combination with BIX-01294 (Cat. No. 3364), helps generate induced pluripotent stem cells (iPSCs) from mouse embryonic fibroblasts (MEFs). Inhibits autophagy.

(S)-(-)-Enantiomer and (R)-(+)-Enantiomer also available.

Specifications

Chemical Identifiers

71145-03-4
356.301
methyl 2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-1,4-dihydropyridine-3-carboxylate, 3-pyridinecarboxylic acid, 1,4-dihydro-2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-, methyl ester, 1,4-dihydro-2,6-dimethyl-5-nitro-4-2′-trifluoromethyl phenyl-3-pyridinecarboxylic acid methyl ester, bay-k-8644, +--isomer, opera_id_789, d01rad, y-bay k 8644, r-+-1,4-dihydro-2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-3-pyridinecarboxylic acid methyl ester
CHEBI:34555
CC1=C(C(C(=C(N1)C)[N+](=O)[O-])C2=CC=CC=C2C(F)(F)F)C(=O)OC
C16H15F3N2O4
ZFLWDHHVRRZMEI-UHFFFAOYSA-N
2303
methyl 2,6-dimethyl-5-nitro-4-[2-(trifluoromethyl)phenyl]-1,4-dihydropyridine-3-carboxylate

Specifications

50mg
71145-03-4
methyl 2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-1,4-dihydropyridine-3-carboxylate, 3-pyridinecarboxylic acid, 1,4-dihydro-2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-, methyl ester, 1,4-dihydro-2,6-dimethyl-5-nitro-4-2′-trifluoromethyl phenyl-3-pyridinecarboxylic acid methyl ester, bay-k-8644, +--isomer, opera_id_789, d01rad, y-bay k 8644, r-+-1,4-dihydro-2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-3-pyridinecarboxylic acid methyl ester
CC1=C(C(C(=C(N1)C)[N+](=O)[O-])C2=CC=CC=C2C(F)(F)F)C(=O)OC
356.301
CHEBI:34555
>99%
(±)-Bay K 8644
C16H15F3N2O4
ZFLWDHHVRRZMEI-UHFFFAOYSA-N
methyl 2,6-dimethyl-5-nitro-4-[2-(trifluoromethyl)phenyl]-1,4-dihydropyridine-3-carboxylate
2303
356.3
Videos
Documents