missing translation for 'onlineSavingsMsg'
Learn More

Bax, clone: 6A7, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-6997-81

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15287967

  • £69.09 / 50µg


Estimated Shipment
01-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The 6A7 antibody reacts with mouse, rat, monkey and human Bax (20 kDa). The 6A7 antibody shows no cross-reaction with Bcl-2 or Bcl-xL proteins. Bcl-2, Bax, and Bcl-xL are members of a protein family that regulates apoptosis; while several family members including Bcl-2 and Bcl-xL inhibit apoptosis, overexpression of Bax promotes programmed cell death. A human Bax protein peptide sequence, aa 12-24, was used as immunogen.Applications Reported: The 6A7 antibody has been reported for use in immunoblotting (WB).Applications Tested: The 6A7 antibody has been tested by Western Blot and can be used at approximately 1 μg/mL. It is recommended that the reagent be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Bax
6A7
Unconjugated
Purified
IgG1
RUO
Primary
Rat
Western Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
50μg
Store at 2-8°C.
Monoclonal
Videos