Learn More
Tocris Bioscience™ Bax channel blocker
Allosteric inhibitor of Bax channel activation
Brand:  Tocris Bioscience™ 216050
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13450694
  • £710.01 / 50mg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
Potent inhibitor of Bax-mediated mitochondrial cytochrome c release (IC50 = 0.52 μM). Suggested to inhibit Bax channel formation/activity.
Specifications
Specifications
Bax channel blocker
>98
50mg
Bax channel blocker
540.12