Learn More
Tocris Bioscience™ Bax channel blocker
Inhibits Bax-mediated mitochondrial cytochrome c release
Brand:  Tocris Bioscience™ 216010
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13440694
  • £189.63 / 10mg
Estimated Shipment date
from Supplier 28-10-2019
Description
Description
Potent inhibitor of Bax-mediated mitochondrial cytochrome c release (IC50 = 0.52 μM). Suggested to inhibit Bax channel formation/activity.
Specifications
Specifications
Bax channel blocker
>98
10mg
Bax channel blocker
540.12