missing translation for 'onlineSavingsMsg'
Learn More

Batimastat, Tocris Bioscience™

Potent, broad spectrum MMP inhibitor

Brand:  RD Systems (Bio-Techne) 29611

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 1mg

 View more versions of this product

Product Code. 13442144

  • £132.30 / 1mg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent, broad spectrum MMP inhibitor (IC50 values are 3, 4, 4, 6 and 20 nM for MMP -1, -2, -9, -7 and -3 respectively). Exhibits antiproliferative, anti-invasive and antimetastatic activity in human ovarian carcinoma xenografts in vivo.
Specifications

Chemical Identifiers

130370-60-4
477.638
batimastat, batimastat bb-94, unii-bk349f52c9, batimastat bb-94, butanediamide, n4-hydroxy-n1-2-methylamino-2-oxo-1-phenylmethyl ethyl-2-2-methylpropyl-3-2-thienylthio methyl-, 2r-1 s* ,2r*,3s*, 2s,3r-5-methyl-3-alphas-alpha-methylcarbamoyl phenethyl carbamoyl-2-2-thienylthio methyl hexanohydroxamic acid, 4-n-hydroxyamino-2r-isobutyl-2s-2-thienylthiomethyl succinyl-l-phenylalanine-n-methylamide, 2r-n-1s-1-benzyl-2-methylamino-2-oxo-ethyl-2-1s-2-hydroxyamino-2-oxo-1-2-thienylsulfanylmethyl ethyl-4-methyl-pentanamide, 2r,3s-n-hydroxy-n'-1s-1-methylcarbamoyl-2-phenylethyl-2-2-methylpropyl-3-thiophen-2-ylsulfanyl methyl butanediamide, butanediamide, n sup 4-hydroxy-n sup 1-2-methylamino-2-oxo-1-phenylmethyl ethyl-2-2-methylpropyl-3-2-thienylthio methyl-, 2r-1 s* ,2r*,3s*
(2S,3R)-N-hydroxy-N'-[(2S)-1-(methylamino)-1-oxo-3-phenylpropan-2-yl]-3-(2-methylpropyl)-2-(thiophen-2-ylsulfanylmethyl)butanediamide
C23H31N3O4S2
XFILPEOLDIKJHX-QYZOEREBSA-N
5362422
CC(C)CC(C(CSC1=CC=CS1)C(=O)NO)C(=O)NC(CC2=CC=CC=C2)C(=O)NC

Specifications

Batimastat
130370-60-4
batimastat, batimastat bb-94, unii-bk349f52c9, batimastat bb-94, butanediamide, n4-hydroxy-n1-2-methylamino-2-oxo-1-phenylmethyl ethyl-2-2-methylpropyl-3-2-thienylthio methyl-, 2r-1 s* ,2r*,3s*, 2s,3r-5-methyl-3-alphas-alpha-methylcarbamoyl phenethyl carbamoyl-2-2-thienylthio methyl hexanohydroxamic acid, 4-n-hydroxyamino-2r-isobutyl-2s-2-thienylthiomethyl succinyl-l-phenylalanine-n-methylamide, 2r-n-1s-1-benzyl-2-methylamino-2-oxo-ethyl-2-1s-2-hydroxyamino-2-oxo-1-2-thienylsulfanylmethyl ethyl-4-methyl-pentanamide, 2r,3s-n-hydroxy-n'-1s-1-methylcarbamoyl-2-phenylethyl-2-2-methylpropyl-3-thiophen-2-ylsulfanyl methyl butanediamide, butanediamide, n sup 4-hydroxy-n sup 1-2-methylamino-2-oxo-1-phenylmethyl ethyl-2-2-methylpropyl-3-2-thienylthio methyl-, 2r-1 s* ,2r*,3s*
CC(C)CC(C(CSC1=CC=CS1)C(=O)NO)C(=O)NC(CC2=CC=CC=C2)C(=O)NC
477.638
477.64
1mg
C23H31N3O4S2
XFILPEOLDIKJHX-QYZOEREBSA-N
(2S,3R)-N-hydroxy-N'-[(2S)-1-(methylamino)-1-oxo-3-phenylpropan-2-yl]-3-(2-methylpropyl)-2-(thiophen-2-ylsulfanylmethyl)butanediamide
5362422
>99%
Videos