missing translation for 'onlineSavingsMsg'
Learn More

Bathophenanthrolinedisulfonic acid, disodium salt hydrate, 97%, Thermo Scientific Chemicals

Catalog Number p-3735549
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
52746-49-3
C24H14N2Na2O6S2
536.48
MFCD00149310
PCNDSIWXTYFWIA-UHFFFAOYSA-L
4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid, disodium salt hydrate
54669730
disodium;4-[7-(4-sulfonatophenyl)-1,10-phenanthrolin-4-yl]benzenesulfonate;hydrate
[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=CC=NC2=C1C=CC1=C2N=CC=C1C1=CC=C(C=C1)S([O-])(=O)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
52746-49-3
C24H14N2Na2O6S2
536.48
MFCD00149310
PCNDSIWXTYFWIA-UHFFFAOYSA-L
4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid, disodium salt hydrate
54669730
disodium;4-[7-(4-sulfonatophenyl)-1,10-phenanthrolin-4-yl]benzenesulfonate;hydrate
[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=CC=NC2=C1C=CC1=C2N=CC=C1C1=CC=C(C=C1)S([O-])(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point >300°C
Color Beige-Pink
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Formula Weight 536.5
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Bathophenanthrolinedisulfonic acid, disodium salt hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'