missing translation for 'onlineSavingsMsg'
Learn More

Basophil Marker Mouse anti-Human, PE, Clone: 2D7, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-9748-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15567226

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

The 2D7 monoclonal antibody recognizes human 2D7 antigen expressed exclusively by human basophils. This human basophil marker is a 7.2-7.5kDa protein present exclusively in the secretory granules of human basophils. Upon activation, basophils may degranulate, thereby decreasing the amount of 2D7 antigen available for staining. This antibody does not stain mast cells.
Specifications

Specifications

Basophil Marker
2D7
PE
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.25μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
2D7
IgG1, kappa
RUO
Primary
Human
Videos
Documents

For Research Use Only.