missing translation for 'onlineSavingsMsg'
Learn More

Basophil Marker Mouse anti-Human, PE, Clone: 2D7, eBioscience™

Mouse Monoclonal Antibody

£72.77 - £186.00

Specifications

Antigen Basophil Marker
Clone 2D7
Target Species Human
Conjugate PE
Isotype IgG1, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15577226 View Documents Affymetrix eBioscience
12-9748-42
100 Tests £186.00
100 tests View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 19-05-2020
Please to purchase this item. Need a web account? Register with us today!
15567226 View Documents Affymetrix eBioscience
12-9748-41
25 Tests This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
Description

Description

The human 2D7 antigen is expressed exclusively by human basophils. This human basophil marker is a 7.2-7.5 kDa protein present exclusively in the secretory granules of human basophils. Upon activation, basophils may degranulate, thereby decreasing the amount of 2D7 antigen available for staining.
Specifications

Specifications

Basophil Marker
Human
IgG1, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
2D7
PE
Flow Cytometry
Primary
Monoclonal
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.