Basophil Marker Mouse anti-Human, eFluor 450, Clone: 2D7, eBioscience™

Mouse Monoclonal Antibody

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: 48-9748-41

Code: NEW

Additional Details:
Additional Details: Weight: 0.23750kg



Disclaimers: For Research Use Only.

Product Code. 15559836

Quantity Price
1 £ 84.99 / 25 tests
Estimated Shipment date
from Supplier 02-05-2017
Add to basket


Description and Specification

Specification

Primary or Secondary Primary
Format Conjugated
Host Species Mouse
Quantity 25 tests
Applications Flow Cytometry (Intracellular Staining)
Isotype IgG1
Species Reactivity Human
Regulatory Status RUO
Conjugate eFluor 450
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Concentration 5μL (0.5μg)/test
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Monoclonal or Polyclonal Monoclonal
Clone 2D7
Gene Alias 2D7
Antigen Basophil Marker

The 2D7 monoclonal antibody recognizes human 2D7 antigen expressed exclusively by human basophils. This human basophil marker is a 7.2-7.5kDa protein present exclusively in the secretory granules of human basophils. Upon activation, basophils may degranulate, thereby decreasing the amount of 2D7 antigen available for staining. This antibody does not stain mast cells.