Basophil Marker Mouse anti-Human, eFluor 450, Clone: 2D7, eBioscience™

Mouse Monoclonal Antibody

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: 48-9748-42

100TEST Anti-Human Basophil Marker eFluor 450

Code: NEW

Additional Details:
Additional Details: Weight: 0.23750kg



Disclaimers: For Research Use Only.

Product Code. 15569836

Quantity Price
1 £ 297.2 / 100 tests
Estimated Shipment date
from Supplier 27-02-2017
Add to basket


Description and Specification

Specification

Isotype IgG1
Regulatory Status RUO
Species Reactivity Human
Clone 2D7
Antigen Basophil Marker
Format Conjugated
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Gene Alias 2D7
Host Species Mouse
Applications Flow Cytometry (Intracellular Staining)
Quantity 100 tests
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 5μL (0.5μg)/test
Conjugate eFluor 450

The 2D7 monoclonal antibody recognizes human 2D7 antigen expressed exclusively by human basophils. This human basophil marker is a 7.2-7.5kDa protein present exclusively in the secretory granules of human basophils. Upon activation, basophils may degranulate, thereby decreasing the amount of 2D7 antigen available for staining. This antibody does not stain mast cells.