Learn More
Thermo Scientific™ Barnstead™ Final Filter
Use the Thermo Scientific™ Barnstead™ Final Filter on select models of Barnstead pure and ultrapure water systems.
Brand:  Thermo Scientific™ D3750
110.10 GBP valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Product Code. 10092323
  • £118.56 / Each
Focus On Clean Water
Use promo code 24139 at checkout
View Available Promotions
Only null left
Estimated Shipment: 07-08-2025
Add to basket
Description
Description
Use the Thermo Scientific™ Barnstead™ Final Filter on select models of Barnstead pure and ultrapure water systems. Compatible with Thermo Scientific™ B-Pure™, Nanopure™, Easypure™ II, Easypure™ II RF, Easypure™ RoDi, E-Pure™ (3,4 holder) and TII systems.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.2µm Final Filter
0.2 μm
B-Pure, Nanopure, Easypure II, Easypure II RF, Easypure RoDi, E-Pure (3,4 holder), and TII systems
Final Filter