missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Barium tungsten isopropoxide, 10% w/v in isopropanol

Thin film, refractories, mixed oxide precursor

Brand:  Alfa Aesar™ 041046.AC

Code : NEW

Additional Details : CAS Number : 333971-25-8 Weight : 0.01000kg Transport : UN number : 1219 Chem class : 1 Pack group : II

Quantity 25mL

 View more versions of this product

Product Code. 11329467

  • £102.40 / 25mL


Estimated Shipment date
from Supplier 21-03-2018
Add to basket

Specifications

Chemical Identifiers

Barium tungsten isopropoxide
BaW[OCH(CH3)2]8
MFCD00798550
333971-25-8
793.88
UN1219

Specifications

Barium tungsten isopropoxide
Liquid
793.88
Moisture Sensitive
Dark brown
25mL
333971-25-8
BaW[OCH(CH3)2]8
MFCD00798550
UN1219
10% w/v in isopropanol
Videos
SDS