missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Barium titanium oxide sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.9% (metals basis excluding Sr)
Brand: Alfa Aesar™ 044806.KS
Code : NEW
Additional Details : Weight : 0.01000kg
Quantity | 1 each |
---|
Specifications
Barium titanium oxide sputtering target | |
99.9% | |
BaTiO3 | |
1 each |
50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick | |
(metals basis excluding Sr) | |
MFCD00003447 |