missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Barium tantalum oxide, 99% (metals basis)

Brand:  Alfa Aesar™ 039179.09

Code : NEW

Additional Details : CAS Number : 12047-34-6 Weight : 0.01000kg Transport : UN number : 1564 Chem class : 1 Pack group : III

Quantity 10g

 View more versions of this product

Product Code. 11373558

  • £55.33 / 10g


Estimated Shipment date
from Supplier 25-05-2018
Add to basket

Specifications

Chemical Identifiers

Barium tantalum oxide
595.2
UN1564
12047-34-6
MFCD00210601

Specifications

Barium tantalum oxide
Powder
(metals basis)
MFCD00210601
−100 Mesh
12047-34-6
99%
595.2
UN1564
10g
Videos
SDS