missing translation for 'onlineSavingsMsg'
Learn More

Barium Sulfate, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog Number p-5390707
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
7727-43-7
BaO4S
233.38
MFCD00003455
TZCXTZWJZNENPQ-UHFFFAOYSA-L
barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
24414
[Ba++].[O-]S([O-])(=O)=O
This item is not returnable. View return policy
7727-43-7
BaO4S
233.38
MFCD00003455
TZCXTZWJZNENPQ-UHFFFAOYSA-L
barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
24414
[Ba++].[O-]S([O-])(=O)=O

High quality chemicals for laboratory and specialized industrial use

Physical Form Powder
Quantity 500 g
IUPAC Name barium(2+);sulfate
Formula Weight 233.39
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 4.50kg
Chemical Name or Material Barium Sulfate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'