missing translation for 'onlineSavingsMsg'
Learn More

Barium Sulfate, Powder, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Brand:  JT Baker 1030-01

Additional Details : CAS Number : 7727-43-7 Weight : 0.65000kg

Packaging Poly Bottle
Quantity 500g

 View more versions of this product

Product Code. 15567884

  • £311.00 / 500g

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

7727-43-7
233.383
barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
barium(2+);sulfate
BaO4S
TZCXTZWJZNENPQ-UHFFFAOYSA-L
24414
[O-]S(=O)(=O)[O-].[Ba+2]

Specifications

Barium Sulfate
BaO4S
TZCXTZWJZNENPQ-UHFFFAOYSA-L
barium(2+);sulfate
24414
Powder
Poly Bottle
500g
7727-43-7
barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
[O-]S(=O)(=O)[O-].[Ba+2]
233.383
233.39
BAKER ANALYZED™ Reagent
1L = 4.50kg
Videos
Documents