Learn More
Thermo Scientific Alfa Aesar Barium rod, 22mm (0.9 in.) dia. x 450mm (17.7 in.), 99+% (metals basis), Sr =0.8%
Brand: Thermo Scientific Alfa Aesar 041302.KF
Barium rod, 22mm (0.9 in.) dia. x 450mm (17.7 in.), 99+% (metals basis), Sr =0.8%
Code : NEW
Additional Details : CAS Number : 7440-39-3 Weight : 0.01000kg Transport : UN number : 1400 Chem class : 4.3 Pack group : II