missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium nitrate, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 592.0°C |
Quantity | 2.5 kg |
Linear Formula | Ba(NO3)2 |
Merck Index | 15,979 |
Solubility Information | Solubility in water: 9 g/100mL (20°C). Other solubilities: practically insoluble in alcohol and acetone |
IUPAC Name | barium(2+);dinitrate |
Formula Weight | 261.34 |
Grade | ACS Reagent |
Packaging | Plastic bottle |
Chemical Name or Material | Barium nitrate |
Safety and Handling
Product Identifier
- Barium nitrate
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- H319-Causes serious eye irritation.
- H301-Toxic if swallowed.
- H332-Harmful if inhaled.
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'