missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Description
Barium nitrate is used by the oil and gas industries to make drilling muds. It is used to make paints, bricks, tiles, glass, rubber, and other barium compounds. It also used in is used in signal flares and water softener.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 592°C (decomposition) |
Color | White |
Physical Form | Crystalline |
Quantity | 5 g |
Assay Percent Range | 99.99% (Metal basis) |
Solubility Information | Soluble in water. Slightly soluble in ethanol and acetone. Partially soluble in cold water. |
IUPAC Name | barium(2+) dinitrate |
Formula Weight | 261.35 |
Sensitivity | Hygroscopic |
Appearance | White crystalline |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'