missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Barium isopropoxide, 99.5% (metals basis)

Brand:  Alfa Aesar™ 041271.03

Code : NEW

Additional Details : CAS Number : 24363-37-9 Weight : 0.00100kg Transport : UN number : 3205 Chem class : 4.2 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 11319657

  • £26.50 / 1g


Estimated Shipment date
from Supplier 22-06-2018
Add to basket

Specifications

Chemical Identifiers

Barium isopropoxide
Ba[OCH(CH3)2]2
MFCD00050486
24363-37-9
255.52
UN3205

Specifications

Barium isopropoxide
Powder/Lump
(metals basis)
255.52
Moisture Sensitive
White
1g
24363-37-9
99.5%
Ba[OCH(CH3)2]2
MFCD00050486
UN3205
≈200°C (dec.)
Videos
SDS